Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008112.4 | NP_032138.3 | 11 | 1536 | ||
GenBank mRNA | AK007871.1 | - | 11 | 1479 | ||
AK078144.1 | - | 1479 | ||||
AK079690.1 | - | 10 | 1377 | |||
AK083063.1 | - | 11 | 1477 | |||
AK090083.1 | - | 11 | 1484 | |||
AK137761.1 | - | 11 | 1483 | |||
AK143633.1 | - | 11 | 1481 | |||
AK143681.1 | - | 11 | 1515 | |||
AK149958.1 | - | 11 | 1489 | |||
AK150356.1 | - | 11 | 1471 | |||
AK150655.1 | - | 11 | 1506 | |||
AK152250.1 | - | 11 | 1470 | |||
AK153381.1 | - | 11 | 1449 | |||
AK159734.1 | - | 11 | 1481 | |||
AK166317.1 | - | 11 | 1481 | |||
AK167659.1 | - | 11 | 1493 | |||
AK169299.1 | - | 11 | 1447 | |||
BC024971.1 | - | 11 | 1457 | |||
BC053381.1 | - | 3636 | ||||
BC055341.1 | - | 11 | 1462 | 6808475 | ||
L36314.1 | - | 11 | 1342 |