Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077705.2 | NP_001071173.1 | 3 | 479 | ||
NM_013545.3 | NP_038573.2 | 3 | 477 | |||
GenBank mRNA | AK132509.1 | - | 3 | 396 | ||
AK150509.1 | - | 3 | 384 | |||
AK151077.1 | - | 3 | 398 | |||
AK151623.1 | - | 3 | 401 | |||
AK159686.1 | - | 3 | 436 | |||
AK159833.1 | - | 3 | 429 | |||
AK167923.1 | - | 3 | 431 | |||
AK169804.1 | - | 3 | 479 | |||
BC012660.1 | - | 3 | 422 | 4010462 | ||
BY747031.1 | - | 3 | 468 | |||
M68902.1 | - | 3 | 429 | |||
M90389.1 | - | 3 | 379 |