Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPA8 SIRNA Assays ›
          Gene Symbol:
          Hspa8
          Gene Name:
          heat shock protein 8
          Gene Aliases:
          2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
          Chromosome Location:
          Chr.9: 40801273 - 40805199 on Build GRCm38
          UniGene ID:
          Mm.290774
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS274956
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hspa8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031165.4 NP_112442.2 3 316
          GenBank mRNA AK004608.1 - 2 605
          AK035286.1 - 3 314
          AK075935.1 - 3 313
          AK081701.1 - 3 314
          AK098074.1 - 3 313
          AK145416.1 - 3 236
          AK145579.1 - 3 310
          AK146547.1 - 3 317
          AK146708.1 - 3 311
          AK146985.1 - 3 314
          AK147864.1 - 3 314
          AK149954.1 - 3 311
          AK150474.1 - 3 308
          AK150498.1 - 3 308
          AK150701.1 - 3 318
          AK150958.1 - 3 311
          AK151041.1 - 3 314
          AK151065.1 - 3 314
          AK151127.1 - 3 317
          AK151287.1 - 3 314
          AK151435.1 - 3 314
          AK151516.1 - 3 320
          AK151537.1 - 3 311
          AK151741.1 - 3 314
          AK151775.1 - 3 316
          AK151808.1 - 3 317
          AK151865.1 - 3 314
          AK151892.1 - 3 316
          AK151948.1 - 3 317
          AK151997.1 - 3 314
          AK152315.1 - 3 317
          AK152598.1 - 3 314
          AK152697.1 - 3 314
          AK152703.1 - 3 314
          AK152768.1 - 3 317
          AK152803.1 - 3 317
          AK153032.1 - 3 316
          AK153834.1 - 3 314
          AK157831.1 - 3 314
          AK159479.1 - 3 314
          AK162775.1 - 3 314
          AK163987.1 - 3 314
          AK164000.1 - 3 314
          AK166643.1 - 3 328
          AK166721.1 - 3 311
          AK166767.1 - 3 314
          AK166776.1 - 3 317
          AK166808.1 - 3 314
          AK166830.1 - 3 312
          AK166846.1 - 3 314
          AK166861.1 - 3 317
          AK166873.1 - 3 314
          AK166908.1 - 3 317
          AK166910.1 - 3 314
          AK166913.1 - 3 314
          AK166933.1 - 3 314
          AK167043.1 - 3 316
          AK167121.1 - 3 317
          AK167122.1 - 3 314
          AK167134.1 - 3 312
          AK167163.1 - 3 316
          AK167218.1 - 3 314
          AK167229.1 - 3 315
          AK167254.1 - 3 315
          AK167414.1 - 3 314
          AK167845.1 - 3 317
          AK167910.1 - 3 317
          AK168435.1 - 3 317
          AK168492.1 - 3 316
          AK168519.1 - 3 314
          AK168542.1 - 3 316
          AK168577.1 - 3 314
          AK168625.1 - 3 314
          AK168654.1 - 3 314
          AK168711.1 - 3 314
          AK168750.1 - 3 317
          AK168776.1 - 3 317
          AK168887.1 - 3 316
          AK168934.1 - 3 317
          AK169093.1 - 3 314
          AK169179.1 - 3 314
          AK169236.1 - 3 314
          AK169293.1 - 3 328
          AK169594.1 - 3 313
          AK171434.1 - 3 315
          AK185576.1 - 2 75
          AK188190.1 - 1 22
          AK190603.1 - 1 22
          AK199049.1 - 1 22
          AK204952.1 - 1 22
          AK206788.1 - 1 22
          AK212540.1 - 1 22
          AK214010.1 - 3 302
          AK218563.1 - 1 22
          AK220016.1 - 1 22
          BC006722.1 - 3 338 3711399
          BC066191.1 - 3 270 30467052
          BC085486.1 - 3 302 30540914
          BC089322.1 - 3 295 30538640
          BC089457.1 - 3 273 30628505
          BC094900.1 - 3 287 30063483
          BC106193.1 - 3 298 6329283
          M19141.1 - 3 300
          U27129.1 - 3 265

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          protein folding
          RNA splicing
          protein refolding
          negative regulation of transcription, DNA-templated
          ATP metabolic process
          positive regulation of mRNA splicing, via spliceosome
          chaperone mediated protein folding requiring cofactor
          regulation of cell cycle
          chaperone-mediated protein folding
          clathrin coat disassembly
          negative regulation of fibril organization
          nucleotide binding
          G-protein coupled receptor binding
          receptor binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          MHC class II protein complex binding
          heat shock protein binding
          ubiquitin protein ligase binding
          ATPase activity, coupled
          ADP binding
          poly(A) RNA binding
          unfolded protein binding
          C3HC4-type RING finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0