Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HSPA8 SIRNA Assays ›
Gene Symbol:
Hspa8
Gene Name:
heat shock protein 8
Gene Aliases:
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
Chromosome Location:
Chr.9: 40801273 - 40805199 on Build GRCm38
UniGene ID:
Mm.290774
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS274957
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hspa8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031165.4
NP_112442.2
4
626
GenBank mRNA
AK004608.1
-
3
915
AK035286.1
-
4
624
AK075935.1
-
4
623
AK081701.1
-
4
625
AK145416.1
-
4
546
AK145579.1
-
4
620
AK146547.1
-
4
627
AK146708.1
-
4
621
AK146985.1
-
4
624
AK147864.1
-
4
624
AK149954.1
-
4
621
AK150474.1
-
4
618
AK150498.1
-
4
618
AK150701.1
-
4
628
AK150958.1
-
4
621
AK151041.1
-
4
624
AK151065.1
-
4
624
AK151127.1
-
4
627
AK151287.1
-
4
624
AK151435.1
-
4
624
AK151516.1
-
4
630
AK151537.1
-
4
621
AK151741.1
-
4
624
AK151775.1
-
4
626
AK151808.1
-
4
627
AK151865.1
-
4
624
AK151892.1
-
4
626
AK151948.1
-
4
627
AK151997.1
-
4
624
AK152315.1
-
4
627
AK152598.1
-
4
624
AK152697.1
-
4
624
AK152703.1
-
4
624
AK152768.1
-
4
627
AK152803.1
-
4
627
AK153032.1
-
4
626
AK153834.1
-
4
624
AK159479.1
-
4
624
AK163987.1
-
4
624
AK164000.1
-
4
624
AK166545.1
-
4
626
AK166643.1
-
4
638
AK166721.1
-
4
621
AK166767.1
-
4
624
AK166776.1
-
4
627
AK166808.1
-
4
624
AK166830.1
-
4
622
AK166846.1
-
4
624
AK166861.1
-
4
627
AK166873.1
-
4
624
AK166908.1
-
4
627
AK166910.1
-
4
624
AK166913.1
-
4
624
AK166933.1
-
4
624
AK167043.1
-
4
626
AK167121.1
-
4
627
AK167122.1
-
4
624
AK167134.1
-
4
622
AK167163.1
-
4
626
AK167218.1
-
4
624
AK167229.1
-
4
625
AK167254.1
-
4
625
AK167414.1
-
4
624
AK168435.1
-
4
627
AK168492.1
-
4
626
AK168511.1
-
2
162
AK168519.1
-
4
624
AK168542.1
-
4
626
AK168577.1
-
4
624
AK168625.1
-
4
624
AK168654.1
-
4
624
AK168711.1
-
4
624
AK168750.1
-
4
627
AK168776.1
-
4
627
AK168887.1
-
4
626
AK168934.1
-
4
627
AK169093.1
-
4
624
AK169179.1
-
4
624
AK169236.1
-
4
624
AK169293.1
-
4
638
AK169329.1
-
4
627
AK171434.1
-
4
625
AK178143.1
-
1
105
AK190509.1
-
1
105
AK193850.1
-
1
105
AK193882.1
-
1
105
AK195240.1
-
1
105
AK198438.1
-
1
105
AK198532.1
-
2
166
AK199049.1
-
2
332
AK199142.1
-
1
105
AK199143.1
-
1
105
AK199178.1
-
1
105
AK199354.1
-
1
105
AK199410.1
-
1
138
AK199504.1
-
1
105
AK200099.1
-
1
105
AK200309.1
-
1
105
AK200579.1
-
1
105
AK201767.1
-
1
105
AK202859.1
-
1
105
AK203911.1
-
1
105
AK205187.1
-
1
105
AK206134.1
-
1
105
AK206953.1
-
1
105
AK212914.1
-
1
125
AK214003.1
-
2
152
BC066191.1
-
4
580
30467052
BC085486.1
-
4
612
30540914
BC089322.1
-
4
605
30538640
BC089457.1
-
4
583
30628505
BC106193.1
-
4
608
6329283
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
protein refolding
negative regulation of transcription, DNA-templated
ATP metabolic process
positive regulation of mRNA splicing, via spliceosome
chaperone mediated protein folding requiring cofactor
regulation of cell cycle
chaperone-mediated protein folding
clathrin coat disassembly
negative regulation of fibril organization
nucleotide binding
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
MHC class II protein complex binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
ADP binding
poly(A) RNA binding
unfolded protein binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0