Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JUN SIRNA Assays ›
          Gene Symbol:
          Jun
          Gene Name:
          jun proto-oncogene
          Gene Aliases:
          AP-1, Junc, c-jun
          Chromosome Location:
          Chr.4: 95049036 - 95052222 on Build GRCm38
          UniGene ID:
          Mm.275071
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS275169
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Jun
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010591.2 NP_034721.1 1 1884
          GenBank mRNA AK133722.1 - 1 1865
          AK133867.1 - 1 1867
          AK135023.1 - 1 1863
          AK140911.1 - 1 1859
          AK159196.1 - 1 1859
          AK159220.1 - 1 1865
          BC002081.1 - 1 1027 3493248
          BC021888.1 - 1 1818 3156091
          BC032086.1 - 1 1352
          BC055887.1 - 1 1961
          BC094032.1 - 2 1746 4936392
          J04115.1 - 1 1849
          X12740.1 - 1 1547
          X12761.1 - 1 1287

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          microglial cell activation
          release of cytochrome c from mitochondria
          liver development
          positive regulation of endothelial cell proliferation
          outflow tract morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          transforming growth factor beta receptor signaling pathway
          SMAD protein import into nucleus
          learning
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to radiation
          cellular process
          monocyte differentiation
          axon regeneration
          negative regulation of protein autophosphorylation
          response to lipopolysaccharide
          cellular response to hormone stimulus
          response to cytokine
          leading edge cell differentiation
          response to muscle stretch
          response to drug
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of DNA binding
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          positive regulation of GTPase activity
          negative regulation by host of viral transcription
          positive regulation by host of viral transcription
          positive regulation of cell differentiation
          positive regulation of monocyte differentiation
          positive regulation of DNA replication
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          cellular response to potassium ion starvation
          response to cAMP
          regulation of cell cycle
          membrane depolarization
          SMAD protein signal transduction
          cellular response to calcium ion
          negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          positive regulation of DNA-templated transcription, initiation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription coactivator activity
          GTPase activator activity
          protein binding
          transcription factor binding
          enzyme binding
          cAMP response element binding
          protein homodimerization activity
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          poly(A) RNA binding
          protein heterodimerization activity
          R-SMAD binding
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline