Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163510.1 | NP_001156982.1 | 7, 8 | 1284 | ||
NM_008433.4 | NP_032459.3 | 7, 8 | 1284 | |||
GenBank mRNA | AB128930.1 | - | 5, 6 | 1047 | ||
AF042487.1 | - | 7, 8 | 1150 | |||
AF072884.1 | - | 7, 8 | 1110 | |||
AK010943.1 | - | 7, 8 | 1285 | |||
AK085752.1 | - | 7, 8 | 1101 | |||
AK088778.1 | - | 2, 3 | 208 | |||
AK155910.1 | - | 7, 8 | 1249 | |||
BC010274.1 | - | 7, 8 | 1233 | 3589411 |