Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145831.1 | NP_001139303.1 | 17 | 2199 | ||
NM_001145832.1 | NP_001139304.1 | 13 | 1713 | |||
NM_010631.2 | NP_034761.3 | 15 | 2056 | |||
GenBank mRNA | AB001457.1 | - | 4 | 345 | ||
AF013118.2 | - | 11 | 1692 | |||
AK132895.1 | - | 13 | 1713 | |||
AK135926.1 | - | 10 | 1255 | |||
AK166482.1 | - | 17 | 2179 | |||
BC004069.1 | - | 7 | 804 | |||
BC016118.1 | - | 10 | 1260 | |||
BC023374.1 | - | 8 | 848 | |||
BC070429.1 | - | 15 | 2056 | 6810893 | ||
D49546.1 | - | 15 | 2071 |
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047800.1 | - | 1702 |