Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290814.1 | NP_001277743.1 | 15 | 3004 | ||
GenBank mRNA | BC052061.1 | - | 15 | 2334 | ||
BU056443.1 | - | 2 | 312 |