Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145953.1 | NP_001139425.1 | 3 | 156 | ||
NM_010705.3 | NP_034835.1 | 3 | 129 | |||
GenBank mRNA | AK089249.1 | - | 3 | 73 | ||
AK149783.1 | - | 3 | 103 | |||
AK155244.1 | - | 3 | 72 | |||
AK155484.1 | - | 3 | 71 | |||
AK159536.1 | - | 3 | 97 | |||
AK170803.1 | - | 3 | 186 | |||
BB853044.1 | - | 3 | 130 | |||
BC132328.1 | - | 3 | 82 | 40130605 | ||
BC138790.1 | - | 3 | 82 | 8861812 | ||
BC145419.1 | - | 3 | 82 | 9053977 | ||
J03723.1 | - | 1 | 29 | |||
X16074.1 | - | 3 | 176 | |||
X16834.1 | - | 3 | 75 |