Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008691.2 | NP_032717.2 | 2, 3 | 1391 | ||
GenBank mRNA | AK018234.1 | - | 2, 3 | 1230 | ||
AK051296.1 | - | 2, 3 | 1234 | |||
AK051696.1 | - | 2, 3 | 1231 | |||
AK080794.1 | - | 2, 3 | 1235 | |||
AK164318.1 | - | 2, 3 | 1234 | |||
AK165041.1 | - | 2, 3 | 1234 | |||
BC052352.1 | - | 2, 3 | 1233 | |||
BC058333.1 | - | 2, 3 | 1224 | |||
BC119602.2 | - | 2, 3 | 1199 | |||
BC128564.1 | - | 2, 3 | 1194 | |||
DQ201636.1 | - | 2, 3 | 1209 | |||
M20481.1 | - | 2, 3 | 237 |