Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011495.2 | NP_035625.2 | 2 | 412 | ||
NM_173169.2 | NP_775261.2 | 2 | 412 | |||
GenBank mRNA | AK006827.1 | - | 2 | 318 | ||
AK045082.1 | - | 2 | 336 | |||
AK137080.1 | - | 2 | 324 | |||
AK137471.1 | - | 2 | 323 | |||
AK164465.1 | - | 2 | 322 | |||
BC026785.1 | - | 2 | 322 | 5150173 | ||
BC057940.1 | - | 2 | 309 | 5323295 | ||
L29479.1 | - | 2 | 314 | |||
L29480.1 | - | 2 | 314 |