Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9530068E07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153117.2 | NP_694757.1 | 2 | 408 | ||
GenBank mRNA | AK007314.1 | - | 2 | 270 | ||
AK012310.1 | - | 2 | 374 | |||
AK079269.1 | - | 2 | 409 | |||
AK135934.1 | - | 2 | 372 | |||
AK144677.1 | - | 2 | 372 | |||
AK145254.1 | - | 2 | 377 | |||
AK147956.1 | - | 2 | 372 | |||
AK150517.1 | - | 2 | 392 | |||
AK154955.1 | - | 2 | 378 | |||
BC006957.1 | - | 2 | 262 | |||
BC034829.1 | - | 2 | 382 | 4482756 |
Gene Symbol: Igk-V4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | L12155.1 | - | 141 |