Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TEK SIRNA Assays ›
          Gene Symbol:
          Tek
          Gene Name:
          endothelial-specific receptor tyrosine kinase
          Gene Aliases:
          AA517024, Cd202b, Hyk, STK1, Tie-2, Tie2
          Chromosome Location:
          Chr.4: 94739289 - 94874976 on Build GRCm38
          UniGene ID:
          Mm.14313
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS278161
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tek
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290549.1 NP_001277478.1 1 388
          NM_001290551.1 NP_001277480.1 1 388
          NM_013690.3 NP_038718.2 1 388
          GenBank mRNA AK053336.1 - 1 365
          BC050824.1 - 1 382 6333576
          D13738.1 - 1 387
          S67051.1 - 1 51
          X67553.1 - 1 159
          X71426.1 - 1 122

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001105737.

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          patterning of blood vessels
          vasculogenesis
          positive regulation of protein phosphorylation
          endothelial cell proliferation
          regulation of endothelial cell proliferation
          sprouting angiogenesis
          positive regulation of cytokine secretion involved in immune response
          protein phosphorylation
          cell-matrix adhesion
          transmembrane receptor protein tyrosine kinase signaling pathway
          heart development
          positive regulation of endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          phosphorylation
          single organismal cell-cell adhesion
          negative regulation of angiogenesis
          peptidyl-tyrosine phosphorylation
          hemopoiesis
          regulation of cell migration
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          response to retinoic acid
          regulation of establishment or maintenance of cell polarity
          positive regulation of peptidyl-serine phosphorylation
          substrate adhesion-dependent cell spreading
          intracellular signal transduction
          positive regulation of protein import into nucleus
          negative regulation of apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          response to estrogen
          regulation of angiogenesis
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          protein autophosphorylation
          Tie signaling pathway
          protein oligomerization
          positive regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          heart trabecula formation
          positive regulation of ERK1 and ERK2 cascade
          regulation of NIK/NF-kappaB signaling
          positive regulation of intracellular signal transduction
          positive regulation of actin cytoskeleton reorganization
          negative regulation of endothelial cell apoptotic process
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0