Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009462.1 | NP_033488.1 | 4 | 500 | ||
GenBank mRNA | AK077453.1 | - | 4 | 521 | ||
AK129083.1 | - | 4 | 508 | |||
AK137015.1 | - | 4 | 522 | |||
AK153675.1 | - | 4 | 992 | |||
AK157970.1 | - | 4 | 524 | |||
AK165156.1 | - | 4 | 520 | |||
AK166243.1 | - | 4 | 517 | |||
AK172443.1 | - | 4 | 522 | |||
BC007134.1 | - | 4 | 502 | 3491297 | ||
D84096.1 | - | 4 | 500 |