Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009536.4 | NP_033562.3 | 2 | 365 | ||
GenBank mRNA | AF483478.1 | - | 2 | 224 | ||
AF483479.1 | - | 2 | 224 | |||
AK075995.1 | - | 2 | 314 | |||
AK135831.1 | - | 2 | 293 | |||
AK135867.1 | - | 2 | 293 | |||
AK169812.1 | - | 2 | 320 | |||
AK192964.1 | - | 1 | 123 | |||
AK212566.1 | - | 1 | 123 | |||
AK214161.1 | - | 1 | 123 | |||
AK218792.1 | - | 1 | 123 | |||
AK220088.1 | - | 1 | 123 | |||
AX775991.1 | - | 2 | 368 | |||
AX775993.1 | - | 1 | 161 | |||
BC058686.1 | - | 2 | 324 | 6411635 | ||
D87663.1 | - | 2 | 252 | |||
Z19599.1 | - | 2 | 307 |