Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177833.1 | NP_001171304.1 | 3, 4 | 630 | ||
NM_001177834.1 | NP_001171305.1 | 3, 4 | 630 | |||
NM_001177835.1 | NP_001171306.1 | 3, 4 | 630 | |||
NM_001177836.1 | NP_001171307.1 | 3, 4 | 630 | |||
NM_001177837.1 | NP_001171308.1 | 3, 4 | 630 | |||
NM_001177838.1 | NP_001171309.1 | 3, 4 | 630 | |||
NM_001177839.1 | NP_001171310.1 | 3, 4 | 630 | |||
NM_145533.2 | NP_663508.1 | 3, 4 | 630 | |||
GenBank mRNA | AF495851.1 | - | 3, 4 | 498 | ||
AF495852.1 | - | 3, 4 | 499 | |||
AF495853.1 | - | 3, 4 | 499 | |||
AJ567473.1 | - | 3, 4 | 549 | |||
AJ567474.1 | - | 3, 4 | 549 | |||
AJ567476.1 | - | 3, 4 | 549 | |||
AJ567477.1 | - | 3, 4 | 549 | |||
AJ567478.1 | - | 3, 4 | 549 | |||
AJ567479.1 | - | 3, 4 | 549 | |||
AJ567480.1 | - | 3, 4 | 549 | |||
AK143129.1 | - | 3, 4 | 604 | |||
AK159275.1 | - | 3, 4 | 557 | |||
AK159372.1 | - | 3, 4 | 557 | |||
AK166350.1 | - | 529 | ||||
BC004831.1 | - | 3, 4 | 549 | 2647695 |