Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042487.1 | NP_001035952.1 | 6 | 1851 | ||
NM_001042488.2 | NP_001035953.1 | 6 | 1150 | |||
NM_001277186.1 | NP_001264115.1 | 12 | 3086 | |||
NM_001277187.1 | NP_001264116.1 | 11 | 3044 | |||
NM_146128.6 | NP_666240.4 | 12 | 3095 | |||
GenBank mRNA | AK122408.1 | - | 10 | 5753 | ||
AK150554.1 | - | 6 | 1071 | |||
AK156212.1 | - | 4 | 3729 | |||
AK195297.1 | - | 2 | 95 | |||
AY243849.2 | - | 10 | 2672 | |||
BC024558.1 | - | 6 | 1255 | |||
BC058948.1 | - | 6 | 1174 | |||
BC085475.1 | - | 6 | 1638 | |||
BC106094.1 | - | 6 | 1233 | |||
BC141110.1 | - | 10 | 2723 | 9055722 | ||
BC145394.1 | - | 10 | 2723 | 9053952 | ||
BC145395.1 | - | 9 | 2681 | 9053953 |