Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172147.1 | NP_001165618.1 | 4 | 445 | ||
NM_001172148.1 | NP_001165619.1 | 3 | 363 | |||
NM_001290630.1 | NP_001277559.1 | 3 | 363 | |||
NM_153586.2 | NP_705814.1 | 3 | 363 | |||
GenBank mRNA | AK040213.1 | - | 3 | 303 | ||
AK077972.1 | - | 3 | 503 | |||
AK080041.1 | - | 1 | 129 | |||
AK084514.1 | - | 3 | 298 | |||
AK084629.1 | - | 3 | 298 | |||
AK141075.1 | - | 3 | 298 | |||
AK163147.1 | - | 2 | 282 | |||
AK164496.1 | - | 4 | 393 | |||
AK166005.1 | - | 3 | 298 | |||
AK166229.1 | - | 311 | ||||
BC037024.1 | - | 3 | 330 | 5368185 |