Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DHRS9 SIRNA Assays ›
          Gene Symbol:
          Dhrs9
          Gene Name:
          dehydrogenase/reductase (SDR family) member 9
          Gene Aliases:
          C730025I08Rik, Rdh15
          Chromosome Location:
          Chr.2: 69380462 - 69403086 on Build GRCm38
          UniGene ID:
          Mm.211655
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS280638
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dhrs9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175512.2 NP_780721.1 2 312
          GenBank mRNA AF361479.1 - 1 97
          AK050180.1 - 2 330
          AK080914.1 - 2 312
          AY936479.1 - 1 84
          BC118029.1 - 2 273 40093681

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          androgen metabolic process
          progesterone metabolic process
          9-cis-retinoic acid biosynthetic process
          oxidation-reduction process
          alcohol dehydrogenase (NAD) activity
          retinol dehydrogenase activity
          oxidoreductase activity
          pinocarveol dehydrogenase activity
          chloral hydrate dehydrogenase activity
          hydroxymethylmethylsilanediol oxidase activity
          1-phenylethanol dehydrogenase activity
          myrtenol dehydrogenase activity
          cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
          3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
          2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
          cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
          citronellol dehydrogenase activity
          naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
          2,4,4-trimethyl-1-pentanol dehydrogenase activity
          2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
          1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
          endosulfan diol dehydrogenase activity
          endosulfan hydroxyether dehydrogenase activity
          3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
          3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
          versicolorin reductase activity
          ketoreductase activity
          testosterone dehydrogenase (NAD+) activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0