Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252081.1 | NP_001239010.1 | 16 | 3356 | ||
NM_001252082.1 | NP_001239011.1 | 17 | 3275 | |||
NM_027996.3 | NP_082272.1 | 16 | 3377 | |||
GenBank mRNA | AB093273.1 | - | 6 | 1453 | ||
AK053265.1 | - | 16 | 3332 | |||
AK147398.1 | - | 16 | 3378 | |||
BC049362.1 | - | 4 | 428 | |||
BC059058.1 | - | 7 | 1197 | |||
BC085161.1 | - | 17 | 2789 | |||
BC151046.1 | - | 16 | 3276 | 9087959 | ||
BC151056.1 | - | 16 | 3276 | 9087969 | ||
BC151173.1 | - | 16 | 3276 | 9088062 | ||
BC172103.1 | - | 16 | 3252 | 9054389 |