Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252081.1 | NP_001239010.1 | 22 | 5040 | ||
NM_001252082.1 | NP_001239011.1 | 23 | 4959 | |||
NM_027996.3 | NP_082272.1 | 22 | 5061 | |||
GenBank mRNA | AB093273.1 | - | 11 | 3277 | ||
AK009454.1 | - | 2 | 482 | |||
AK147398.1 | - | 22 | 5062 | |||
BC026504.1 | - | 1 | 810 | |||
BC027155.1 | - | 6 | 1513 | |||
BC030914.1 | - | 4 | 1838 | |||
BC037627.1 | - | 3 | 843 | |||
BC049362.1 | - | 10 | 2112 | |||
BC058666.1 | - | 8 | 1318 | |||
BC059058.1 | - | 13 | 2881 | |||
BC085161.1 | - | 23 | 4473 | |||
BC151046.1 | - | 22 | 4960 | 9087959 | ||
BC151056.1 | - | 22 | 4960 | 9087969 | ||
BC151173.1 | - | 22 | 4960 | 9088062 | ||
BC172103.1 | - | 22 | 4936 | 9054389 |