Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166033.1 | NP_001159505.1 | 6 | 2655 | ||
NM_172977.3 | NP_766565.2 | 5 | 2549 | |||
GenBank mRNA | AK030351.1 | - | 5 | 2476 | ||
AK047698.1 | - | 4 | 1659 | |||
AK141269.1 | - | 5 | 2531 | |||
AK146401.1 | - | 6 | 2611 | |||
AK146403.1 | - | 6 | 2611 | |||
AK150650.1 | - | 5 | 2531 | |||
AK151179.1 | - | 5 | 2531 | |||
AK153102.1 | - | 5 | 2531 | |||
BC095998.1 | - | 5 | 2488 | 30736671 | ||
BC117919.1 | - | 5 | 2461 | 40097451 | ||
BC117920.1 | - | 5 | 2459 | 40097456 |