Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam131c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085513.2 | NP_001078982.1 | 5 | 453 | ||
GenBank mRNA | BC147048.1 | - | 5 | 453 | 9056076 | |
BC147049.1 | - | 5 | 453 | 9056071 |