Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdspl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290991.1 | NP_001277920.1 | 13 | 2006 | ||
NM_001290992.1 | NP_001277921.1 | 13 | 2005 | |||
NM_212450.4 | NP_997615.1 | 13 | 2009 | |||
GenBank mRNA | AK028251.1 | - | 11 | 1069 | ||
AK030085.1 | - | 13 | 1525 | |||
AK033382.1 | - | 13 | 1558 | |||
AK035067.1 | - | 2 | 120 | |||
AK044805.1 | - | 13 | 1540 | |||
AK049463.1 | - | 13 | 1525 | |||
AK053513.1 | - | 13 | 1535 | |||
AK082646.1 | - | 13 | 1865 | |||
AK163262.1 | - | 13 | 2010 | |||
BC052660.1 | - | 13 | 1492 | 30049400 | ||
BC066219.1 | - | 13 | 1526 |