Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253857.1 | NP_001240786.1 | 3 | 1929 | ||
NM_027384.1 | NP_081660.1 | 3 | 1929 | |||
GenBank mRNA | AK129421.1 | - | 3 | 182 | ||
AK141438.1 | - | 3 | 195 | |||
CF897091.1 | - | 1 | 577 | |||
GU079948.1 | - | 3 | 1929 |