Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111289.1 | NP_001104759.1 | 11, 12 | 1433 | ||
NM_001111290.1 | NP_001104760.1 | 11, 12 | 1406 | |||
NM_001111291.1 | NP_001104761.1 | 12, 13 | 1473 | |||
NM_001111292.1 | NP_001104762.1 | 11, 12 | 1433 | |||
NM_016739.3 | NP_058019.2 | 12, 13 | 1473 | |||
GenBank mRNA | AB373955.1 | - | 11, 12 | 1295 | ||
AK133891.1 | - | 12, 13 | 1436 | |||
AK141992.1 | - | 11, 12 | 1375 | |||
AK157707.1 | - | 12, 13 | 1457 | |||
AK161131.1 | - | 6, 7 | 568 | |||
AK166461.1 | - | 11, 12 | 1391 | |||
AK168676.1 | - | 11, 12 | 1401 | |||
AK169366.1 | - | 3, 4 | 204 | |||
AK170830.1 | - | 12, 13 | 1435 | |||
BC052427.1 | - | 12, 13 | 1452 | 6836041 | ||
BC127030.1 | - | 3, 4 | 299 | |||
U18773.1 | - | 1, 2 | 94 | |||
U27838.1 | - | 11, 12 | 1410 |