Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205236.1 | NP_001192165.1 | 13, 14 | 2580 | ||
NM_133715.5 | NP_598476.2 | 11, 12 | 1549 | |||
GenBank mRNA | AK046763.1 | - | 11, 12 | 1550 | ||
AK143916.1 | - | 9, 10 | 1021 | |||
AK154926.1 | - | 12, 13 | 2871 |