Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081549.2 | NP_001075018.1 | 2 | 378 | ||
NM_019466.4 | NP_062339.2 | 2 | 224 | |||
GenBank mRNA | AF237789.1 | - | 2 | 128 | ||
AF237790.1 | - | 2 | 128 | |||
AF260717.1 | - | 2 | 142 | |||
AF263239.1 | - | 2 | 158 | |||
AF263240.1 | - | 2 | 241 | |||
AF282255.1 | - | 2 | 135 | |||
AK010696.1 | - | 95 | ||||
AK077283.1 | - | 2 | 224 | |||
AK146764.1 | - | 2 | 353 | |||
AY325904.1 | - | 2 | 334 | |||
BC013551.1 | - | 2 | 204 | 4236038 |