Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164733.1 | NP_001158205.1 | 5 | 653 | ||
NM_001164734.1 | NP_001158206.1 | 5 | 663 | |||
NM_019939.2 | NP_064323.2 | 5 | 663 | |||
GenBank mRNA | AF161181.1 | - | 5 | 527 | ||
AF199009.1 | - | 5 | 659 | |||
AF199010.1 | - | 5 | 659 | |||
AK144434.1 | - | 5 | 664 | |||
AK164549.1 | - | 5 | 654 | |||
BC042868.2 | - | 5 | 610 | |||
BC046421.1 | - | 5 | 610 | |||
BC069908.1 | - | 5 | 652 | |||
BC106140.1 | - | 5 | 610 | |||
BC138666.1 | - | 5 | 521 | 8861687 | ||
BC145363.1 | - | 5 | 521 | 9053921 | ||
BC145364.1 | - | 5 | 511 | 9053922 |