Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145960.1 | NP_001139432.1 | 15 | 1641 | ||
NM_020258.4 | NP_064654.3 | 15 | 1641 | |||
GenBank mRNA | AF121081.1 | - | 15 | 1540 | ||
AK028984.1 | - | 15 | 1317 | |||
AK030881.1 | - | 3 | 850 | |||
AK031045.1 | - | 15 | 1579 | |||
AK079414.1 | - | 15 | 1317 | |||
AK079823.1 | - | 15 | 1642 | |||
AK220151.1 | - | 13 | 1083 | |||
BC022752.1 | - | 1 | 16 | |||
BC038000.1 | - | 15 | 1312 |