Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199275.2 | NP_001186204.1 | 7 | 806 | ||
NM_001199276.2 | NP_001186205.1 | 5 | 476 | |||
NM_001271455.1 | NP_001258384.1 | 6 | 529 | |||
NM_001271456.1 | NP_001258385.1 | 7 | 701 | |||
NM_021327.4 | NP_067302.2 | 6 | 648 | |||
GenBank mRNA | AJ242777.1 | - | 5 | 448 | ||
AJ242778.1 | - | 6 | 620 | |||
AK135338.1 | - | 6 | 627 | |||
AK156131.1 | - | 6 | 501 | |||
AK170841.1 | - | 5 | 454 | |||
AK172890.1 | - | 434 | ||||
AK199662.1 | - | 3 | 169 | |||
AK199664.1 | - | 3 | 169 | |||
AY012159.2 | - | 3 | 218 | |||
BC008186.1 | - | 6 | 565 | 3589311 | ||
BC008665.1 | - | 7 | 672 | 3598934 |