Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290764.1 | NP_001277693.1 | 14, 15 | 1469 | ||
NM_001290766.1 | NP_001277695.1 | 12, 13 | 1453 | |||
NM_001290767.1 | NP_001277696.1 | 12, 13 | 1224 | |||
NM_021549.3 | NP_067524.2 | 13, 14 | 1561 | |||
GenBank mRNA | AF129451.1 | - | 12, 13 | 1219 | ||
AK049992.1 | - | 13, 14 | 1364 | |||
AK159535.1 | - | 13, 14 | 1326 | |||
AK159770.1 | - | 13, 14 | 1326 | |||
AK160020.1 | - | 13, 14 | 1326 | |||
AK160160.1 | - | 14, 15 | 1466 | |||
AK172558.1 | - | 12, 13 | 1258 | |||
BC047269.1 | - | 11, 12 | 1503 | |||
BC057659.1 | - | 13, 14 | 1363 | 2865792 |