Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290764.1 | NP_001277693.1 | 4 | 413 | ||
NM_001290766.1 | NP_001277695.1 | 3 | 505 | |||
NM_001290767.1 | NP_001277696.1 | 3 | 248 | |||
NM_021549.3 | NP_067524.2 | 3 | 505 | |||
GenBank mRNA | AF129451.1 | - | 3 | 247 | ||
AK049992.1 | - | 3 | 308 | |||
AK159535.1 | - | 3 | 271 | |||
AK159770.1 | - | 3 | 271 | |||
AK160020.1 | - | 3 | 271 | |||
AK160160.1 | - | 4 | 409 | |||
AK172558.1 | - | 3 | 310 | |||
AK186762.1 | - | 3 | 118 | |||
BC047269.1 | - | 2 | 555 | |||
BC057659.1 | - | 3 | 307 | 2865792 | ||
CK628081.1 | - | 3 | 317 | |||
CX223963.1 | - | 3 | 492 |