Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025824.3 | NP_080100.1 | 3 | 185 | ||
GenBank mRNA | AK004784.2 | - | 3 | 185 | ||
AK013980.1 | - | 3 | 179 | |||
AK029030.1 | - | 3 | 196 | |||
AK076086.1 | - | 3 | 186 | |||
AK076560.1 | - | 3 | 186 | |||
AK087980.1 | - | 3 | 173 | |||
AK088322.1 | - | 3 | 193 | |||
AK133973.1 | - | 3 | 186 | |||
AK151919.1 | - | 3 | 173 | |||
AK152939.1 | - | 3 | 173 | |||
AK153538.1 | - | 3 | 174 | |||
AK161192.1 | - | 3 | 183 | |||
AK166247.1 | - | 3 | 190 | |||
AK167056.1 | - | 3 | 170 | |||
AK167074.1 | - | 3 | 157 | |||
AK167514.1 | - | 3 | 145 | |||
AK167982.1 | - | 3 | 191 | |||
AK168158.1 | - | 3 | 197 | |||
AK187482.1 | - | 3 | 148 | |||
AK192968.1 | - | 3 | 150 | |||
AK195624.1 | - | 3 | 150 | |||
AK201364.1 | - | 241 | ||||
AK205016.1 | - | 3 | 180 | |||
AK205108.1 | - | 3 | 150 | |||
AK212098.1 | - | 3 | 150 | |||
BC005466.1 | - | 3 | 194 | 3601190 | ||
BC028865.1 | - | 3 | 173 | 4910833 |