Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025824.3 | NP_080100.1 | 6 | 535 | ||
GenBank mRNA | AK004784.2 | - | 6 | 535 | ||
AK013980.1 | - | 6 | 529 | |||
AK029030.1 | - | 6 | 546 | |||
AK076086.1 | - | 6 | 536 | |||
AK076560.1 | - | 6 | 536 | |||
AK087980.1 | - | 6 | 523 | |||
AK088322.1 | - | 6 | 543 | |||
AK133973.1 | - | 6 | 536 | |||
AK151919.1 | - | 6 | 523 | |||
AK152939.1 | - | 6 | 523 | |||
AK153538.1 | - | 6 | 524 | |||
AK161192.1 | - | 6 | 533 | |||
AK166247.1 | - | 6 | 540 | |||
AK167056.1 | - | 6 | 520 | |||
AK167074.1 | - | 6 | 507 | |||
AK167514.1 | - | 6 | 495 | |||
AK167982.1 | - | 6 | 541 | |||
AK168158.1 | - | 6 | 547 | |||
AK198677.1 | - | 3 | 151 | |||
AK215707.1 | - | 2 | 123 | |||
BC005466.1 | - | 6 | 544 | 3601190 | ||
BC028865.1 | - | 6 | 523 | 4910833 |