Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025824.3 | NP_080100.1 | 8 | 809 | ||
GenBank mRNA | AK004784.2 | - | 8 | 809 | ||
AK013980.1 | - | 8 | 803 | |||
AK029030.1 | - | 8 | 820 | |||
AK076086.1 | - | 8 | 810 | |||
AK076560.1 | - | 8 | 810 | |||
AK087980.1 | - | 8 | 797 | |||
AK088322.1 | - | 8 | 817 | |||
AK133973.1 | - | 8 | 810 | |||
AK151919.1 | - | 8 | 797 | |||
AK152939.1 | - | 8 | 796 | |||
AK153538.1 | - | 8 | 798 | |||
AK161192.1 | - | 8 | 807 | |||
AK166247.1 | - | 8 | 814 | |||
AK167056.1 | - | 8 | 794 | |||
AK167074.1 | - | 8 | 781 | |||
AK167514.1 | - | 8 | 768 | |||
AK167982.1 | - | 8 | 815 | |||
AK168158.1 | - | 8 | 821 | |||
AK191609.1 | - | 1 | 12 | |||
AK193767.1 | - | 1 | 12 | |||
AK197959.1 | - | 1 | 12 | |||
AK198435.1 | - | 1 | 12 | |||
AK204270.1 | - | 1 | 12 | |||
BC005466.1 | - | 8 | 818 | 3601190 | ||
BC028865.1 | - | 8 | 797 | 4910833 |