Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284332.1 | NP_001271261.1 | 6 | 805 | ||
NM_025858.3 | NP_080134.1 | 6 | 802 | |||
NM_026381.5 | NP_080657.1 | 4 | 502 | |||
GenBank mRNA | AF520699.1 | - | 6 | 784 | ||
AK009225.1 | - | 6 | 742 | |||
AK018291.1 | - | 4 | 458 | |||
AK020877.1 | - | 4 | 479 | |||
AK033545.1 | - | 6 | 762 | |||
AK081060.1 | - | 4 | 480 | |||
AK088188.1 | - | 5 | 498 | |||
AK090357.1 | - | 4 | 656 | |||
AK090373.1 | - | 6 | 741 | |||
AK150460.1 | - | 5 | 960 | |||
AK151632.1 | - | 5 | 960 | |||
AK183004.1 | - | 2 | 52 | |||
AK184395.1 | - | 2 | 51 | |||
BC010238.1 | - | 6 | 766 | 3581921 | ||
BC017600.1 | - | 4 | 414 | 4487600 | ||
BC057089.1 | - | 4 | 454 | 5689110 |