Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026647.3 | NP_080923.2 | 9 | 1094 | ||
GenBank mRNA | AK020293.1 | - | 9 | 976 | ||
AK144108.1 | - | 8 | 856 | |||
AY166674.1 | - | 7 | 732 | |||
BC043110.1 | - | 9 | 940 | 6406930 |