Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033142.2 | NP_001028314.1 | 2 | 294 | ||
GenBank mRNA | AK079883.1 | - | 2 | 275 | ||
AK151813.1 | - | 2 | 294 | |||
AK165700.1 | - | 1 | 1078 | |||
AK210531.1 | - | 1 | 93 | |||
BC131924.1 | - | 2 | 293 | 40130201 | ||
BC132281.1 | - | 2 | 293 | 40130558 |
Gene Symbol: Gm42051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079889.1 | - | 1 | 57 |