Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290390.1 | NP_001277319.1 | 20, 21 | 3360 | ||
NM_001290392.1 | NP_001277321.1 | 17, 18 | 3095 | |||
NM_001290393.1 | NP_001277322.1 | 19, 20 | 3309 | |||
NM_027671.5 | NP_081947.2 | 20, 21 | 3363 | |||
GenBank mRNA | AF367759.1 | - | 19, 20 | 2015 | ||
AK016927.1 | - | 6, 7 | 652 | |||
AK038698.1 | - | 17, 18 | 3096 | |||
AK080034.1 | - | 20, 21 | 2725 | |||
AK083241.1 | - | 20, 21 | 2348 |