Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028044.2 | NP_082320.1 | 1, 2 | 289 | ||
GenBank mRNA | AK005460.1 | - | 1, 2 | 291 | ||
AK051121.1 | - | 1, 2 | 286 | |||
AK051628.1 | - | 1, 2 | 307 | |||
AK081672.1 | - | 1, 2 | 205 | |||
AK159555.1 | - | 1, 2 | 334 | |||
AK159871.1 | - | 1, 2 | 319 | |||
AK166578.1 | - | 1, 2 | 297 | |||
AK167201.1 | - | 1, 2 | 310 | |||
AK167613.1 | - | 1, 2 | 307 | |||
AK169021.1 | - | 1, 2 | 314 | |||
AK169098.1 | - | 1, 2 | 334 | |||
AK169410.1 | - | 1, 2 | 264 | |||
AK179383.1 | - | 1, 2 | 20 | |||
AK180041.1 | - | 39 | ||||
AK184231.1 | - | 1, 2 | 25 | |||
AK190584.1 | - | 34 | ||||
AK216318.1 | - | 1, 2 | 20 | |||
BC055711.1 | - | 1, 2 | 290 | 6415164 | ||
BC085268.1 | - | 1, 2 | 67 | 6487559 |