Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028044.2 | NP_082320.1 | 2, 3 | 403 | ||
GenBank mRNA | AK005460.1 | - | 2, 3 | 405 | ||
AK051628.1 | - | 2, 3 | 421 | |||
AK081672.1 | - | 2, 3 | 319 | |||
AK159555.1 | - | 2, 3 | 448 | |||
AK159871.1 | - | 2, 3 | 433 | |||
AK166578.1 | - | 2, 3 | 411 | |||
AK167201.1 | - | 2, 3 | 424 | |||
AK167613.1 | - | 2, 3 | 421 | |||
AK169021.1 | - | 2, 3 | 428 | |||
AK169098.1 | - | 2, 3 | 448 | |||
AK169410.1 | - | 2, 3 | 378 | |||
AK179383.1 | - | 2, 3 | 134 | |||
AK180041.1 | - | 1, 2 | 153 | |||
AK184231.1 | - | 2, 3 | 139 | |||
AK208457.1 | - | 1, 2 | 103 | |||
AK212516.1 | - | 1, 2 | 52 | |||
AK215736.1 | - | 1, 2 | 113 | |||
AK216318.1 | - | 2, 3 | 134 | |||
BC055711.1 | - | 2, 3 | 404 | 6415164 | ||
BC085268.1 | - | 2, 3 | 181 | 6487559 |