Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1600014C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085385.1 | NP_001078854.1 | 2 | 330 | ||
NM_028166.3 | NP_082442.1 | 2 | 138 | |||
GenBank mRNA | AK005449.1 | - | 2 | 110 | ||
AK032135.1 | - | 2 | 108 | |||
AK032342.1 | - | 2 | 109 | |||
AK146474.1 | - | 2 | 118 | |||
BC010476.1 | - | 2 | 94 | 3673971 | ||
BC019834.1 | - | 2 | 261 | 5123719 | ||
BC085480.1 | - | 2 | 76 | 6401538 | ||
CK335181.1 | - | 2 | 90 |