Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028572.1 | NP_082848.1 | 2 | 151 | ||
GenBank mRNA | BC042696.1 | - | 2 | 76 |