Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102436.1 | NP_001095906.1 | 10, 11 | 1441 | ||
NM_001102437.1 | NP_001095907.1 | 10, 11 | 1571 | |||
NM_001102438.1 | NP_001095908.1 | 10, 11 | 1568 | |||
NM_028793.3 | NP_083069.1 | 10, 11 | 1444 | |||
GenBank mRNA | AK005001.2 | - | 9, 10 | 1360 | ||
AK050450.1 | - | 10, 11 | 1407 | |||
AK147839.1 | - | 10, 11 | 1571 | |||
AK214423.1 | - | 1, 2 | 125 | |||
BC035202.1 | - | 10, 11 | 1497 | |||
BC053518.1 | - | 10, 11 | 1535 | |||
BC061484.1 | - | 9, 10 | 1342 | 5355466 | ||
BC084584.1 | - | 10, 11 | 1373 | 5257773 | ||
BC090641.1 | - | 10, 11 | 1395 |