Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smlr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195596.1 | NP_001182525.1 | 1 | 254 | ||
GenBank mRNA | AK008073.1 | - | 1 | 252 | ||
AK080199.1 | - | 1 | 126 |