Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253681.1 | NP_001240610.1 | 2, 3 | 272 | ||
NM_001253686.1 | NP_001240615.1 | 2, 3 | 272 | |||
NM_001253687.1 | NP_001240616.1 | 2, 3 | 272 | |||
NM_001253688.1 | NP_001240617.1 | 2, 3 | 272 | |||
NM_001253689.1 | NP_001240618.1 | 2, 3 | 272 | |||
NM_029673.3 | NP_083949.2 | 2, 3 | 272 | |||
GenBank mRNA | AK030841.1 | - | 2, 3 | 214 | ||
AK033126.1 | - | 2, 3 | 183 | |||
AK038129.1 | - | 2, 3 | 262 | |||
AK049189.1 | - | 2, 3 | 216 | |||
AK146640.1 | - | 2, 3 | 264 | |||
AK152588.1 | - | 2, 3 | 218 | |||
AK157805.1 | - | 2, 3 | 213 | |||
AK159909.1 | - | 2, 3 | 223 | |||
AK163491.1 | - | 2, 3 | 180 | |||
AK169365.1 | - | 2, 3 | 175 | |||
AK201051.1 | - | 2, 3 | 155 | |||
CJ058627.1 | - | 2, 3 | 273 |