Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027456.1 | NP_081732.1 | 18, 19 | 1780 | ||
NM_030184.2 | NP_084460.1 | 18, 19 | 1815 | |||
GenBank mRNA | AK017564.1 | - | 18, 19 | 1781 | ||
AK019600.1 | - | 18, 19 | 1804 | |||
AK030307.1 | - | 18, 19 | 1812 | |||
AK039800.1 | - | 18, 19 | 1788 | |||
AK045699.1 | - | 18, 19 | 1779 | |||
AK046693.1 | - | 18, 19 | 1798 | |||
AK076239.1 | - | 1, 2 | 46 | |||
AK140598.1 | - | 18, 19 | 1811 | |||
AK161393.1 | - | 18, 19 | 1795 | |||
AK164041.1 | - | 18, 19 | 1814 | |||
AK186122.1 | - | 1, 2 | 31 | |||
BC028964.1 | - | 11, 12 | 1035 | 5364429 | ||
BC043101.1 | - | 18, 19 | 1767 | 6406321 | ||
BQ173978.1 | - | 3, 4 | 395 |