Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163750.1 | NP_001157222.1 | 6 | 1072 | ||
NM_001163751.1 | NP_001157223.1 | 6 | 727 | |||
NM_027141.2 | NP_081417.2 | 6 | 748 | |||
GenBank mRNA | AF403038.1 | - | 6 | 748 | ||
AK009325.1 | - | 6 | 748 | |||
AK076170.1 | - | 6 | 1024 | |||
AK082768.1 | - | 6 | 671 | |||
AK156579.1 | - | 5 | 1085 | |||
AK167474.1 | - | 5 | 1065 | |||
AK176934.1 | - | 2 | 87 | |||
AK183595.1 | - | 2 | 87 | |||
AK191468.1 | - | 2 | 87 | |||
AK208055.1 | - | 2 | 87 | |||
AK208775.1 | - | 2 | 91 | |||
AK210660.1 | - | 2 | 87 | |||
AK212011.1 | - | 1 | 162 | |||
AK212237.1 | - | 1 | 162 | |||
AK214702.1 | - | 1 | 162 | |||
AK220234.1 | - | 6 | 1010 | |||
BC026523.1 | - | 6 | 1413 | |||
BC072556.1 | - | 6 | 1033 | 5691114 |
Gene Symbol: Eme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC034637.1 | - | 2665 |