Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053716.1 | NP_446168.1 | 6 | 863 | ||
GenBank mRNA | AJ005046.1 | - | 6 | 863 |